siRNA Details
Gene Symbol: Snhg11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175692.3 | NP_783623.2 | 2, 3 | 495 | ||
GenBank mRNA | AK080745.1 | - | 2, 3 | 453 | ||
BC030414.1 | - | 2, 3 | 411 | 5366924 | ||
BC049241.1 | - | 409 | 4505955 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
