siRNA Details
Gene Symbol: Ubald2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176902.3 | NP_795876.1 | 2 | 353 | ||
GenBank mRNA | AK050732.1 | - | 2 | 235 | ||
AK144901.1 | - | 2 | 355 | |||
BC116983.1 | - | 2 | 224 | 40126302 | ||
BC119080.1 | - | 2 | 225 | 8733829 | ||
BY734584.1 | - | 2 | 355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
