siRNA Details
Gene Symbol: Hs3st5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081208.2 | NP_001074677.1 | 4 | 1485 | ||
NM_001253355.1 | NP_001240284.1 | 6 | 1649 | |||
NM_001253356.1 | NP_001240285.1 | 5 | 1590 | |||
GenBank mRNA | AK031910.1 | - | 6 | 1639 | ||
BC079867.1 | - | 4 | 1266 | |||
BC147490.1 | - | 6 | 1642 | 9056285 | ||
BC147493.1 | - | 5 | 1583 | 9056309 | ||
BC150973.1 | - | 6 | 1642 | 9087886 | ||
BC158004.1 | - | 6 | 1642 | 9088149 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
