siRNA Details
Gene Symbol: Serpine3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199945.1 | NP_001186874.1 | 3 | 356 | ||
GenBank mRNA | AK053602.1 | - | 3 | 358 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Serpine3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199945.1 | NP_001186874.1 | 3 | 356 | ||
GenBank mRNA | AK053602.1 | - | 3 | 358 |