siRNA Details
Gene Symbol: Ppm1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110218.1 | NP_001103688.1 | 9 | 1760 | ||
NM_176919.4 | NP_795893.2 | 9 | 1760 | |||
GenBank mRNA | AK040194.1 | - | 9 | 1504 | ||
AK040207.1 | - | 9 | 1745 | |||
AK051434.1 | - | 9 | 1719 | |||
AK134804.1 | - | 9 | 1740 | |||
AK150309.1 | - | 9 | 1322 | |||
AK220260.1 | - | 9 | 1717 | |||
BC052910.1 | - | 9 | 1071 |
