siRNA Details
Gene Symbol: 6030419C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176921.3 | NP_795895.1 | 3 | 1389 | ||
GenBank mRNA | AK031382.1 | - | 3 | 1391 | ||
BC096614.1 | - | 2 | 813 | 6468423 |

More Information
Other Species Targeted

Gene Ontology Categories:
