siRNA Details
Gene Symbol: Cxxc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004367.4 | NP_001004367.2 | 3 | 1515 | ||
GenBank mRNA | AK034523.1 | - | 1 | 61 | ||
BC065149.1 | - | 4 | 1201 | |||
BC067052.1 | - | 3 | 1045 | 30619087 | ||
BC082332.1 | - | 2 | 732 | 30537505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
