siRNA Details
Gene Symbol: Cxxc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004367.4 | NP_001004367.2 | 3, 4 | 1575 | ||
GenBank mRNA | BC065149.1 | - | 4, 5 | 1261 | ||
BC067052.1 | - | 3, 4 | 1105 | 30619087 | ||
BC082332.1 | - | 2, 3 | 792 | 30537505 |

More Information
Other Species Targeted

Gene Ontology Categories:
