siRNA Details
Gene Symbol: Pced1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114541.1 | NP_001108013.1 | 7 | 1237 | ||
NM_178762.4 | NP_848877.2 | 6 | 1824 | |||
GenBank mRNA | AK031469.1 | - | 3 | 2955 | ||
AK032354.1 | - | 6 | 1816 | |||
AK032762.1 | - | 6 | 1799 | |||
AK044590.1 | - | 3 | 2521 | |||
AK045248.1 | - | 5 | 1894 | |||
AK085613.1 | - | 5 | 1884 | |||
AK180923.1 | - | 1 | 239 | |||
AK194074.1 | - | 1 | 58 | |||
AK213870.1 | - | 1 | 240 | |||
BC064802.1 | - | 7 | 1283 | 30009464 |

More Information
Other Species Targeted

Gene Ontology Categories:
