siRNA Details
Gene Symbol: Dusp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176933.4 | NP_795907.1 | 3, 4 | 928 | ||
GenBank mRNA | AK053746.1 | - | 3, 4 | 925 | ||
AK080964.1 | - | 3, 4 | 928 | |||
AK165991.1 | - | 3, 4 | 1244 | |||
AK180902.1 | - | 1, 2 | 232 | |||
BC118941.1 | - | 3, 4 | 899 | 40092218 | ||
BC118942.1 | - | 3, 4 | 899 | 40092219 |
