siRNA Details
Gene Symbol: B230216G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242345.1 | NP_001229274.1 | 5, 6 | 315 | ||
GenBank mRNA | AK043509.1 | - | 3, 4 | 228 | ||
AK045625.1 | - | 2, 3 | 409 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
