siRNA Details
Gene Symbol: 9630028B13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036028.1 | - | 2 | 730 | ||
AK048735.1 | - | 2 | 808 | |||
BC147657.1 | - | 2 | 708 | 9087446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
