siRNA Details
Gene Symbol: Celf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176954.3 | NP_795928.2 | 12 | 1289 | ||
GenBank mRNA | AK029796.1 | - | 12 | 2463 | ||
AK132321.1 | - | 12 | 1255 | |||
AK158669.1 | - | 11 | 1066 | |||
BC079845.1 | - | 12 | 1260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
