siRNA Details
Gene Symbol: Zfp653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177318.2 | NP_796292.1 | 5 | 1301 | ||
GenBank mRNA | AK046492.1 | - | 4 | 1348 | ||
AK089104.1 | - | 5 | 1113 | |||
AK157635.1 | - | 4 | 1348 | |||
AY072705.1 | - | 5 | 1311 | |||
BC038037.1 | - | 5 | 1301 | 4190337 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
