siRNA Details
Gene Symbol: Fam168a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178764.3 | NP_848879.1 | 5 | 494 | ||
GenBank mRNA | AK043083.1 | - | 5 | 500 | ||
AK043356.1 | - | 4 | 437 | |||
AK047153.1 | - | 5 | 464 | |||
AK048559.1 | - | 4 | 437 | |||
AK051453.1 | - | 4 | 437 | |||
AK084931.1 | - | 4 | 386 | |||
AK122235.1 | - | 5 | 531 | |||
AK137424.1 | - | 5 | 379 | |||
AK162512.1 | - | 4 | 494 | |||
BC076580.1 | - | 5 | 494 | 30542683 | ||
BC079886.1 | - | 5 | 494 | 6855858 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
