siRNA Details
Gene Symbol: Sybu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032727.1 | NP_001027899.1 | 6 | 1836 | ||
NM_001285840.1 | NP_001272769.1 | 7 | 1942 | |||
NM_001285841.1 | NP_001272770.1 | 4 | 1183 | |||
NM_001285842.1 | NP_001272771.1 | 5 | 1327 | |||
NM_001285843.1 | NP_001272772.1 | 6 | 1394 | |||
NM_001285844.1 | NP_001272773.1 | 5 | 1286 | |||
NM_176998.4 | NP_795972.2 | 8 | 2143 | |||
NM_178765.3 | NP_848880.2 | 8 | 2040 | |||
GenBank mRNA | AB232447.1 | - | 8 | 2143 | ||
AB232448.1 | - | 6 | 1836 | |||
AB232449.1 | - | 8 | 2040 | |||
AK029072.1 | - | 7 | 1639 | |||
AK030482.1 | - | 8 | 1772 | |||
AK030720.1 | - | 9 | 1772 | |||
AK163432.1 | - | 8 | 1805 | |||
AK220268.1 | - | 7 | 1942 | |||
BC048945.1 | - | 6 | 1568 | 6467420 | ||
BC065045.1 | - | 8 | 2037 | 5686153 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
