siRNA Details
Gene Symbol: Zfp944 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176962.4 | NP_795936.2 | 6 | 1731 | ||
GenBank mRNA | AK031839.1 | - | 7 | 1570 | ||
BC108345.1 | - | 7 | 1747 | 4935739 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
