siRNA Details
Gene Symbol: Dcp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033379.3 | NP_001028551.2 | 9 | 1714 | ||
GenBank mRNA | AK047334.1 | - | 9 | 1570 | ||
AK049193.1 | - | 9 | 1642 | |||
AK050548.1 | - | 9 | 1580 | |||
AK053525.1 | - | 2 | 452 | |||
AK153860.1 | - | 9 | 1716 | |||
BC068119.1 | - | 6 | 4020 | |||
BC141162.1 | - | 9 | 1693 | 9055774 | ||
BC141163.1 | - | 9 | 1693 | 9055775 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
