siRNA Details
Gene Symbol: Itpripl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033380.3 | NP_001028552.1 | 1 | 936 | ||
GenBank mRNA | AK134593.1 | - | 1 | 884 | ||
AK137678.1 | - | 1 | 938 | |||
AK149729.1 | - | 1 | 850 | |||
BC150856.1 | - | 1 | 587 | 9087767 | ||
BC150864.1 | - | 1 | 587 | 9087775 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
