siRNA Details
Gene Symbol: Fsd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195284.1 | NP_001182213.1 | 4 | 440 | ||
NM_176966.4 | NP_795940.1 | 4 | 440 | |||
GenBank mRNA | AK038893.1 | - | 4 | 418 | ||
AK052914.1 | - | 4 | 483 | |||
AK140020.1 | - | 4 | 424 | |||
BB662298.1 | - | 4 | 478 | |||
BC130265.1 | - | 4 | 356 | 40142990 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
