siRNA Details
Gene Symbol: Rab9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176971.2 | NP_795945.1 | 3 | 332 | ||
GenBank mRNA | AB232603.1 | - | 1 | 70 | ||
AK034442.1 | - | 3 | 333 | |||
AK049693.1 | - | 3 | 359 | |||
AK161867.1 | - | 3 | 549 | |||
BC107383.2 | - | 3 | 304 | 40057619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
