siRNA Details
Gene Symbol: Mpzl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093749.2 | NP_001087218.1 | 2 | 195 | ||
GenBank mRNA | AK041037.1 | - | 2 | 189 | ||
AK049878.1 | - | 2 | 187 | |||
AK155483.1 | - | 2 | 124 | |||
BC132150.1 | - | 2 | 178 | |||
EF102773.1 | - | 2 | 159 |
