siRNA Details
Gene Symbol: Mpzl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093749.2 | NP_001087218.1 | 2 | 171 | ||
GenBank mRNA | AK041037.1 | - | 2 | 165 | ||
AK049878.1 | - | 2 | 163 | |||
AK155483.1 | - | 2 | 100 | |||
BC132150.1 | - | 2 | 154 | |||
EF102773.1 | - | 2 | 135 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
