siRNA Details
Gene Symbol: Zfp865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033383.2 | NP_001028555.1 | 2 | 560 | ||
NM_001290426.1 | NP_001277355.1 | 2 | 560 | |||
GenBank mRNA | AK051004.1 | - | 2 | 366 | ||
AK154737.1 | - | 2 | 561 | |||
BC020373.1 | - | 2 | 351 |
