siRNA Details
Gene Symbol: Tmem72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178768.4 | NP_848883.2 | 5 | 792 | ||
GenBank mRNA | AK033856.1 | - | 5 | 928 | ||
AK049048.1 | - | 5 | 947 | |||
AK049938.1 | - | 5 | 905 | |||
AK085603.1 | - | 5 | 793 | |||
AK141578.1 | - | 5 | 949 | |||
BC093486.1 | - | 5 | 935 | 30697044 | ||
BC116440.1 | - | 5 | 776 | 40104615 | ||
BC116441.1 | - | 5 | 776 | 40104616 |
