siRNA Details
Gene Symbol: Tmem72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178768.4 | NP_848883.2 | 5 | 843 | ||
GenBank mRNA | AK033856.1 | - | 5 | 979 | ||
AK049048.1 | - | 5 | 998 | |||
AK049938.1 | - | 5 | 956 | |||
AK085603.1 | - | 5 | 844 | |||
AK141578.1 | - | 5 | 1000 | |||
BC093486.1 | - | 5 | 986 | 30697044 | ||
BC116440.1 | - | 5 | 827 | 40104615 | ||
BC116441.1 | - | 5 | 827 | 40104616 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
