siRNA Details
Gene Symbol: Nwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177006.3 | NP_795980.2 | 7 | 4978 | ||
GenBank mRNA | AB093290.1 | - | 7 | 3846 | ||
AK136869.1 | - | 1 | 777 | |||
BC062655.1 | - | 1 | 2468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
