siRNA Details
Gene Symbol: Ccdc79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180958.3 | NP_851289.2 | 4 | 329 | ||
GenBank mRNA | AB775896.1 | - | 2 | 110 | ||
AK029754.1 | - | 4 | 330 | |||
BC113771.1 | - | 3 | 653 | |||
BC129814.1 | - | 3 | 654 |
