siRNA Details
Gene Symbol: Relt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177073.6 | NP_796047.2 | 4 | 401 | ||
GenBank mRNA | AK042363.1 | - | 4 | 396 | ||
AK049073.1 | - | 4 | 402 | |||
AK088621.1 | - | 4 | 384 | |||
BC100310.1 | - | 4 | 391 | 30839974 | ||
BC131929.1 | - | 4 | 312 | 40130206 | ||
BC131931.1 | - | 4 | 312 | 40130208 | ||
BC145200.1 | - | 4 | 312 | 9053751 |
