siRNA Details
Gene Symbol: Fbxl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199632.1 | NP_001186561.1 | 21 | 2469 | ||
NM_177076.4 | NP_796050.2 | 20 | 2370 | |||
GenBank mRNA | AK029566.1 | - | 20 | 2372 | ||
BC145291.1 | - | 21 | 2395 | 9053845 | ||
BC145292.1 | - | 20 | 2297 | 9053846 | ||
CO797010.1 | - | 4 | 489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
