siRNA Details
Gene Symbol: B430306N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177083.4 | NP_796057.1 | 5 | 853 | ||
GenBank mRNA | AK036709.1 | - | 5 | 854 | ||
AK046672.1 | - | 854 | ||||
AK133242.1 | - | 5 | 1714 | |||
AK155881.1 | - | 5 | 869 | |||
AK170621.1 | - | 5 | 875 | |||
AK171230.1 | - | 5 | 2532 | |||
AK171587.1 | - | 5 | 854 | |||
AK171603.1 | - | 5 | 852 | |||
AK171605.1 | - | 5 | 869 | |||
AY522648.1 | - | 5 | 854 | |||
DQ087179.1 | - | 5 | 606 | |||
DQ087180.1 | - | 6 | 697 |
