siRNA Details
Gene Symbol: Zdhhc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172554.2 | NP_766142.2 | 8, 9 | 942 | ||
GenBank mRNA | AK038670.1 | - | 8, 9 | 886 | ||
AK039575.1 | - | 8, 9 | 943 | |||
AK046891.1 | - | 8, 9 | 884 | |||
AK122406.1 | - | 8, 9 | 1016 | |||
AK134967.1 | - | 8, 9 | 891 | |||
BC027101.1 | - | 8, 9 | 830 | |||
BC058772.1 | - | 8, 9 | 883 | |||
BC080664.1 | - | 8, 9 | 881 | |||
BC117788.1 | - | 8, 9 | 884 | 40092137 | ||
BC117789.2 | - | 8, 9 | 884 | 40092140 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001039340, AY886533.
Gene Ontology Categories:
