siRNA Details
Gene Symbol: PTPRJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002843.3 | NP_002834.3 | 11 | 2682 | ||
XM_017018083.1 | XP_016873572.1 | 10 | 2413 | |||
XM_017018084.1 | XP_016873573.1 | 11 | 2795 | |||
XM_017018085.1 | XP_016873574.1 | 11 | 2294 | |||
GenBank mRNA | D37781.1 | - | 11 | 2671 | ||
U10886.1 | - | 11 | 2676 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
