siRNA Details
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045001.1 | - | 1 | 1079 | ||
AK080905.1 | - | 1 | 906 | |||
AK086551.1 | - | 1 | 857 | |||
AK087678.1 | - | 2 | 941 | |||
BC150968.1 | - | 1 | 815 | 9087880 |
