siRNA Details
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045001.1 | - | 1 | 1075 | ||
AK080905.1 | - | 1 | 902 | |||
AK086551.1 | - | 1 | 853 | |||
AK087678.1 | - | 2 | 937 | |||
BC150968.1 | - | 1 | 811 | 9087880 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
