siRNA Details
Gene Symbol: Ttll5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081423.2 | NP_001074892.2 | 17 | 1662 | ||
GenBank mRNA | AB093278.1 | - | 17 | 1595 | ||
AK009255.1 | - | 1 | 42 | |||
AM690748.1 | - | 16 | 1427 | |||
BC035276.1 | - | 16 | 1537 |
