siRNA Details
Gene Symbol: D130009I18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046103.1 | - | 255 | |||
AK051169.1 | - | 1 | 261 | |||
AK051225.1 | - | 1 | 262 | |||
AK051495.1 | - | 1 | 241 | |||
AK051771.1 | - | 1 | 262 |
