siRNA Details
Gene Symbol: Iqcb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177128.4 | NP_796102.2 | 4 | 406 | ||
GenBank mRNA | AK078554.1 | - | 4 | 399 | ||
AK165220.1 | - | 4 | 390 | |||
BC029084.1 | - | 4 | 346 | 3157011 |

More Information
Other Species Targeted

Gene Ontology Categories:
