siRNA Details
Gene Symbol: Rnf152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160368.1 | NP_001153840.1 | 2 | 1930 | ||
NM_178779.4 | NP_848894.1 | 3 | 2053 | |||
GenBank mRNA | AK044638.1 | - | 3 | 2012 | ||
AK081452.1 | - | 1 | 81 | |||
BC118961.2 | - | 2 | 1674 | 40094497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
