siRNA Details
Gene Symbol: Rnf152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160368.1 | NP_001153840.1 | 2 | 570 | ||
NM_178779.4 | NP_848894.1 | 3 | 693 | |||
GenBank mRNA | AK035832.1 | - | 3 | 489 | ||
AK044638.1 | - | 3 | 654 | |||
BC118961.2 | - | 2 | 316 | 40094497 | ||
CJ112559.1 | - | 2 | 570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
