siRNA Details
Gene Symbol: Lypd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177139.5 | NP_796113.1 | 4 | 456 | ||
GenBank mRNA | AK046251.1 | - | 1 | 12 | ||
AK053620.1 | - | 5 | 587 | |||
BB619054.1 | - | 4 | 423 | |||
BB653059.1 | - | 4 | 417 | |||
BC070462.1 | - | 4 | 477 | 30650880 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
