siRNA Details
Gene Symbol: Tmem251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177140.3 | NP_796114.1 | 2 | 630 | ||
GenBank mRNA | AK032513.1 | - | 2 | 636 | ||
AK052027.1 | - | 2 | 632 | |||
AK133919.1 | - | 2 | 633 | |||
AK144149.1 | - | 633 | ||||
BC096606.1 | - | 2 | 618 | 5366962 |
