siRNA Details
Gene Symbol: Itpkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081175.1 | NP_001074644.1 | 2 | 1281 | ||
GenBank mRNA | AK046612.1 | - | 2 | 1231 | ||
AK053759.1 | - | 2 | 1230 | |||
BC151160.1 | - | 2 | 1147 | 9088049 | ||
BC157986.1 | - | 2 | 1146 | 9088142 | ||
BC158000.1 | - | 2 | 1146 | 9088148 |
