siRNA Details
Gene Symbol: Gchfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177157.4 | NP_796131.1 | 3 | 205 | ||
GenBank mRNA | AK028116.1 | - | 3 | 206 | ||
AK028122.1 | - | 3 | 206 | |||
BC096406.1 | - | 3 | 193 | 1511037 | ||
BC132450.1 | - | 3 | 201 | 40130727 | ||
BC132478.1 | - | 3 | 201 | 40130755 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
