siRNA Details
Gene Symbol: Rinl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177158.5 | NP_796132.2 | 8 | 840 | ||
GenBank mRNA | AK030993.1 | - | 8 | 833 | ||
AK082828.1 | - | 1 | 76 | |||
BC044761.1 | - | 8 | 876 | |||
BC046530.1 | - | 8 | 876 | 5329601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
