siRNA Details
Gene Symbol: Rinl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177158.5 | NP_796132.2 | 8 | 843 | ||
GenBank mRNA | AK030993.1 | - | 8 | 836 | ||
AK082828.1 | - | 1 | 79 | |||
BC044761.1 | - | 8 | 879 | |||
BC046530.1 | - | 8 | 879 | 5329601 |
