siRNA Details
Gene Symbol: Rinl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177158.5 | NP_796132.2 | 8 | 1112 | ||
GenBank mRNA | AK030993.1 | - | 8 | 1105 | ||
AK082828.1 | - | 1 | 348 | |||
AK162469.1 | - | 1 | 17 | |||
AK168472.1 | - | 1 | 275 | |||
BC044761.1 | - | 8 | 1148 | |||
BC046530.1 | - | 8 | 1148 | 5329601 |
