siRNA Details
Gene Symbol: Vwc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177164.3 | NP_796138.2 | 2 | 1118 | ||
GenBank mRNA | AB374230.1 | - | 1 | 314 | ||
AK080585.1 | - | 4 | 567 | |||
AK083856.1 | - | 2 | 1083 | |||
BC064448.1 | - | 2 | 931 | |||
BC094541.1 | - | 2 | 1118 | 6410144 | ||
EF552208.1 | - | 1 | 382 | |||
EF552209.1 | - | 1 | 317 | |||
EF552210.1 | - | 2 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
