siRNA Details
Gene Symbol: Heatr5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081179.1 | NP_001074648.1 | 9 | 1407 | ||
GenBank mRNA | AK053037.1 | - | 9 | 1391 | ||
AK079542.1 | - | 9 | 1352 | |||
AK146166.1 | - | 9 | 1389 | |||
AK158478.1 | - | 9 | 1392 | |||
AK164172.1 | - | 2 | 281 | |||
AK220390.1 | - | 6 | 940 | |||
BC086652.1 | - | 8 | 1343 | 6856641 |
