siRNA Details
Gene Symbol: Heatr5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081179.1 | NP_001074648.1 | 6 | 818 | ||
GenBank mRNA | AK053037.1 | - | 6 | 802 | ||
AK079542.1 | - | 6 | 763 | |||
AK158478.1 | - | 6 | 803 | |||
AK220390.1 | - | 3 | 351 | |||
BC086652.1 | - | 5 | 754 | 6856641 |
